GW21280
Anti-CSRP2 antibody produced in chicken
affinity isolated antibody, buffered aqueous solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
biological source
chicken
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
mouse, rat, human
manufacturer/tradename
Genway 15-288-21280
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... CSRP2(1466)
Immunogen
Immunogen Sequence: GI # 4503101, sequence 50-125
Recombinant cysteine and glycine-rich protein 2
Application
Anti-CSRP2 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.
Biochem/physiol Actions
Cysteine and glycine-rich protein 2 is a protein encoded by the CSRP2 gene in humans and belongs to CRP family. It is mapped to chromosome subband 12q21.1 and this region is affected by deletion or breakage events in various tumor types. CSRP2 may have a function in controlling cell growth and differentiation.
Physical form
Solution in phosphate buffered saline containing 0.02% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
R Weiskirchen et al.
The Journal of biological chemistry, 270(48), 28946-28954 (1995-12-01)
Here we describe a family of closely related LIM domain proteins in avian cells. The LIM motif defines a zinc-binding domain that is found in a variety of transcriptional regulators, proto-oncogene products, and proteins associated with sites of cell-substratum contact.
M K Jain et al.
The Journal of biological chemistry, 271(17), 10194-10199 (1996-04-26)
Differentiated, quiescent vascular smooth muscle cells assume a dedifferentiated, proliferative phenotype in response to injury, one of the hallmarks of arteriosclerosis. Members of the LIM family of zinc-finger proteins are important in the differentiation of various cells including striated muscle.
R Weiskirchen et al.
Genomics, 44(1), 83-93 (1997-08-15)
The CSRP2 gene encoding the LIM domain protein CRP2 was originally identified in quail based on its strong transcriptional suppression in transformed avian fibroblasts. Here we have isolated a human CSRP2 cDNA clone encoding a 193-amino-acid human CRP2 (hCRP2) protein
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service