Skip to Content
Merck
All Photos(1)

Documents

GE17-1181-01

RESOURCE RPC

Cytiva 17-1181-01, pack of 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000
NACRES:
NA.56

ligand

matrix surface

feature

autoclavable 20 min at 121°C

packaging

pack of 1 mL

manufacturer/tradename

Cytiva 17-1181-01

parameter

flow rate
580 psi

bed size

6.4 mm × 30 mm

bed volume

1 mL

matrix

polystyrene/divinylbenzene

avg. part. size

15 μm (monosized)

cleaning

1-14

working range

1-12

capacity

~10 mg binding capacity

suitability

suitable for bioprocess medium

General description

RESOURCE RPC are prepacked SOURCE 15RPC columns designed for rapid screening experiments, method development, and small-scale protein purification using reversed phase chromatography.
~10 mg BSA /ml medium at 300 cm/h, ~30 mg bacitracin/ml medium at 300 cm/h, ~50 mg insulin/ml medium at 300 cm/h

Application

SOURCE 15RPC is based on rigid, monosized 15 μm diameter polystyrene/divinyl benzene beads. The matrix has outstanding selectivity for RPC. The monodispersity of the beads yields stable beds, Low back pressures and excellent results at high flow rates.With its high physical and chemical stability and very high batch-to-batch reproducibility, SOURCE 15RPC is well suited for all stages of an industrial scale operation - from research and process development through scale-up and into production. SOURCE 15RPC offers properties superior to those of other polymeric matrices.

Features and Benefits

  • Wide pH range (1-12), outstanding selectivity, chemical resistance, high capacity, and high resolution at high flow rates.
  • Excellent scalability, from RESOURCE to FineLINE columns.
  • using ÄKTATM design and other high-performance liquid chromatography systems.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol or 70% Acetonitrile)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

RESOURCE is a trademark of Cytiva
SOURCE is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service