Skip to Content
Merck
All Photos(1)

Key Documents

F9181

Sigma-Aldrich

Anti-FRAT1 (C-terminal region) antibody produced in rabbit

~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Frequently rearranged in advanced T-cell lymphomas, Anti-GBP, Anti-GSK-3 binding protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~29 kDa

species reactivity

human

concentration

~1.5 mg/mL

technique(s)

western blot: 1-2 μg/mL using HEK-293T cell lysate expressing human FRAT1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... FRAT1(10023)

General description

FRAT1 (frequently rearranged in advanced T-cell lymphomas-1) is a GSK3β binding protein consisting of a conserved GSK3β interacting domain. In human two FRAT genes have been identified, FRAT1 and FRAT2 whereas FRAT1-3 have been identified in mouse.

Application

Anti-FRAT1 (C-terminal region) antibody is suitable for western blot at a concentration of 1-2μg/mL using rat liver extract (S1 fraction) and a HEK-293T cell lysate expressing human FRAT1.

Biochem/physiol Actions

By inhibiting GSK-3-mediated phosphorylation of β-catenin, FRAT1 plays a major role in the wnt-signaling pathway. During binding to the GSKβ3, it competes with axin and thus displacing GSKβ3 from the axin-β-catenin complex. In the canonical wnt signaling pathway, FRAT1 acts more efficiently than FRAT2. FRAT1 has correlation with clinicopathologic features. It has been reported that in several human malignant tumors, FRAT is overexpressed. It′s upregulated expression have been found in several human cancer lines such as gastric cancers and esophageal squamous cell carcinoma (ESCC).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah J Freemantle et al.
Gene, 291(1-2), 17-27 (2002-07-04)
We have isolated the entire coding sequence of human FRAT2 (frequently rearranged in advanced T-cell lymphomas-2). It exhibits appreciable amino acid identity to FRAT1 (77%) which was initially isolated as frequently being overexpressed in a murine leukemia virus insertion model
Yong Zhang et al.
Virchows Archiv : an international journal of pathology, 459(3), 255-263 (2011-08-06)
Frat1 has been reported to be overexpressed in several human malignant tumors, including esophageal squamous, cervical, breast, and ovarian carcinoma, but the role of Frat1 in lung cancer is unknown. Our purpose is to investigate the expression of Frat1 and
Tetsuroh Saitoh et al.
International journal of oncology, 20(4), 785-789 (2002-03-15)
FRAT1 and FRAT2 genes, clustered in human chromosome 10q24, are human homologues to mouse proto-oncogene Frat1, which promotes carcinogenesis through activation of the WNT - beta-catenin - TCF signaling pathway. FRAT1 and FRAT2 mRNAs are up-regulated together in a gastric
Yihua Wang et al.
International journal of cancer, 123(3), 561-568 (2008-05-24)
Esophageal squamous cell carcinoma (ESCC) is an aggressive tumor with a poor prognosis. Although aberrant activation of beta-catenin/T-cell factor (TCF) pathway has been observed in ESCC, mechanisms underlying this phenomenon remain unknown. Frequently rearranged in advanced T-cell lymphomas-1 (FRAT1), overexpressed
Masaru Katoh
Current drug targets, 9(7), 565-570 (2008-08-05)
WNT family members are secreted-type glycoproteins to orchestrate embryogenesis, to maintain homeostasis, and to induce pathological conditions. FZD1, FZD2, FZD3, FZD4, FZD5, FZD6, FZD7, FZD8, FZD9, FZD10, LRP5, LRP6, and ROR2 are transmembrane receptors transducing WNT signals based on ligand-dependent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service