Skip to Content
Merck
All Photos(1)

Documents

EP0030119843

Eppendorf® Tube Rack for cryogenic Tubes

Holds 36 x cryotubes, 3 rows of 12 wells each, pkg of 2 pieces, white, autoclavable

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.12

material

polypropylene

feature

autoclavable
3 rows of 12 wells each
Holds 36 x cryotubes

packaging

pkg of 2 pieces

manufacturer/tradename

Eppendorf® 30119843

technique(s)

cryopreservation: suitable

color

white

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service