Skip to Content
Merck
All Photos(1)

Key Documents

EHU138791

Sigma-Aldrich

MISSION® esiRNA

targeting human POU4F1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACTCAGCCAGAGCACCATCTGCAGGTTCGAGTCGCTCACGCTCTCGCACAACAACATGATCGCGCTCAAGCCCATCCTGCAGGCGTGGCTCGAGGAGGCCGAGGGCGCCCAGCGCGAGAAAATGAACAAGCCTGAGCTCTTCAACGGCGGCGAGAAGAAGCGCAAGCGGACTTCCATCGCCGCGCCCGAGAAGCGCTCCCTCGAGGCCTACTTCGCCGTGCAGCCCCGGCCCTCGTCCGAGAAGATCGCCGCCATCGCCGAGAAACTGGACCTCAAAAAGAACGTGGTGCGGGTGTGGTTTTGCAACCAGAGACAGAAGCAGAAGCGGATGAAATTCTCTGCCACTTACTGAGGGGGCTGGGAGGTGTCGGGCGGGACAGAATGGGGAGCTGAGGAGGCATTTTTGGGGGGCTTTCCTCTGCTTGCCTCCCCTCGGATTTGGAGTGTCCGTTATCCTGCCTGCATTTGGGGAGTCCCTTCTCGCTCTCTTTCCTCCACCCATTCTCTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seung-Nam Jung et al.
The Journal of clinical endocrinology and metabolism, 105(9) (2020-06-01)
Brn3a/Pou4f1 is a class IV POU domain-containing transcription factor and has been found to be expressed in a variety of cancers. However, the mechanism and action of Brn3a in thyroid cancer has not been investigated. To investigate the role of
Lin Liu et al.
Cell death & disease, 11(6), 451-451 (2020-06-14)
BRAF inhibitors (BRAFi) have shown remarkable clinical efficacy in the treatment of melanoma with BRAF mutation. Nevertheless, most patients end up with the development of BRAFi resistance, which strongly limits the clinical application of these agents. POU4F1 is a stem

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service