Skip to Content
Merck
All Photos(1)

Key Documents

EHU047751

Sigma-Aldrich

MISSION® esiRNA

targeting human CX3CL1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCACCTTCTGCCATCTGACTGTCCTGCTGGCTGGACAGCACCACGGTGTGACGAAATGCAACATCACGTGCAGCAAGATGACATCAAAGATACCTGTAGCTTTGCTCATCCACTATCAACAGAACCAGGCATCATGCGGCAAACGCGCAATCATCTTGGAGACGAGACAGCACAGGCTGTTCTGTGCCGACCCGAAGGAGCAATGGGTCAAGGACGCGATGCAGCATCTGGACCGCCAGGCTGCTGCCCTAACTCGAAATGGCGGCACCTTCGAGAAGCAGATCGGCGAGGTGAAGCCCAGGACCACCCCTGCCGCCGGGGGAATGGACGAGTCTGTGGTCCTGGAGCCCGAAGCCACAGGCGAAAGCAGTAGCCTGGAGCCGACTCCTTCTTCCCAGGAAGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ju-Fang Liu et al.
Oncotarget, 8(33), 54136-54148 (2017-09-15)
Osteosarcoma is the most common primary bone tumor in children and teens. The exact molecular mechanism underlying osteosarcoma progression still remains unclear. The CX3CL1/fractalkine has been implicated in various tumors but not in osteosarcoma. This study is the first to
Claudia Geismann et al.
International journal of molecular sciences, 19(6) (2018-06-06)
Pancreatic ductal adenocarcinoma (PDAC) is one of the most lethal malignant neoplasms and registers rising death rates in western countries. Due to its late detection in advanced stages, its extremely aggressive nature and the minimal effectiveness of currently available therapies
Feifei Ren et al.
Immunology and cell biology, 97(5), 457-469 (2018-12-24)
Mutations in the isocitrate dehydrogenase (IDH) 1 gene, especially the R132H mutation, have been reported to be associated with a better prognosis in glioma patients. However, the underlying molecular mechanisms are not yet well understood. Many factors may contribute to
Monika Siwetz et al.
The American journal of pathology, 185(5), 1334-1343 (2015-03-15)
The pathogenesis of preeclampsia (PE) includes the release of placental factors into the maternal circulation, inducing an inflammatory environment in the mother. One of the factors may be the proinflammatory chemokine fractalkine, which is expressed in the syncytiotrophoblast of human
Monika Siwetz et al.
Histochemistry and cell biology, 143(6), 565-574 (2015-01-09)
The chemokine fractalkine (CX3CL1) recently attracted increasing attention in the field of placenta research due to its dual nature, acting both as membrane-bound and soluble forms. While the membrane-bound form mediates flow-resistant adhesion of leukocytes to endothelial and epithelial cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service