Skip to Content
Merck
All Photos(1)

Key Documents

E3401

Sigma-Aldrich

Anti-Elk-1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

western blot (chemiluminescent): 1:1,000 using Elk-1 fusion protein

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ELK1(2002)

Specificity

The antibody detects total Elk-1 independent of phosphorylation state.

Immunogen

synthetic peptide corresponding to amino acid residues 379-392 of human Elk-1.

Physical form

Solution in 10 mM sodium HEPES, pH 7.5, containing 150 mM sodium chloride, 100 μg/ml bovine serum albumin and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fumiaki Uchiumi et al.
Oncology reports, 44(1), 283-292 (2020-05-08)
trans‑Resveratrol (Rsv) is a natural compound contained in red wine and grape skins that has various beneficial effects for organisms such as lengthening of their life span. Rsv induces expression of the human TP53 and HELB genes, which are involved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service