D8136
Aldrithiol™-4
Synonym(s):
4,4′-Dipyridyl disulfide, Aldrithiol-4
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
EC Number:
UNSPSC Code:
12352103
Recommended Products
storage temp.
2-8°C
Legal Information
Aldrithiol is a trademark of Sigma-Aldrich Co. LLC
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service