Skip to Content
Merck
All Photos(1)

Documents

D5179

Sigma-Aldrich

3′-Deoxycytidine

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N3O4
CAS Number:
Molecular Weight:
227.22
MDL number:
UNSPSC Code:
12352204

Assay

≥95% (HPLC)

impurities

≤5% 2′-deoxycytidine

SMILES string

NC1=NC(=O)N(C=C1)C2OC(CO)CC2O

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott A Gabel et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 13(2), 207-217 (2007-10-25)
Phosphate esters play a central role in cellular energetics, biochemical activation, signal transduction and conformational switching. The structural homology of the borate anion with phosphate, combined with its ability to spontaneously esterify hydroxyl groups, suggested that phosphate ester recognition sites
Robert W King et al.
Antiviral chemistry & chemotherapy, 13(6), 363-370 (2003-04-30)
The development of low molecular weight inhibitors of hepatitis C virus (HCV) replication has been hindered by the lack of a good cell-based system that models the entire HCV replication cycle. To date the only two therapies approved for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service