Skip to Content
Merck
All Photos(1)

Documents

CLS431132

Corning® Roller Bottle Caps

vented Easy Grip polyethylene Roller Bottle Cap, individually wrapped, sterile, 300/cs

Synonym(s):

Corning roller bottles, replacement roller bottle caps, roller bottle caps, roller bottles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122105
NACRES:
NB.13

material

high-density polyethylene cap (easy grip)

description

Easy Grip, vented cap

sterility

sterile; γ-irradiated

feature

disposable

packaging

pack of 1
case of 300 (individually wrapped)

manufacturer/tradename

Corning 431132

color

red cap

Looking for similar products? Visit Product Comparison Guide

General description

  • Features large knurls for ergonomic handling
  • Contains hydrophobic membrane for applications requiring gas exchange
  • Sterilized by gamma radiation
  • Does not fit 490cm2 roller bottles

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service