CLS3724BC
Corning® 1536 well plate, with generic barcode
size 1536 wells, non-treated, 50/cs, flat bottom black polystyrene wells, non-sterile, lid: no
Synonym(s):
1536 multiwell plate, 1536 well barcode plate, 1536 well microplate, 1536 well microtiter plate, 1536 well plate, 1536 well plate with generic barcode
About This Item
Recommended Products
material
black plate
flat bottom black polystyrene wells
polystyrene plate
sterility
non-sterile
feature
lid: no
packaging
pack of 10
case of 50
manufacturer/tradename
Corning 3724BC
maximum volume
12.5 μL
size
1536 wells
working volume
8 μL
color
black
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Corning barcode labels have been quality tested for optimal readability, chemical resistance and temperature variation.
- Resistant to commonly used laboratory reagents (100% DMSO, Methanol, Ethanol, Ethyl Acetate, Methylene Chloride, Isopropanol, Acetronitrile, Ethanol/Water/TFA, Sodium Hydroxide, Hydrochloric Acid)
- Resistant to temperature ranges of -80ºC to +52ºC
Technical Details
- Bar Code labeling specifications
- 2.75" x 0.25" label size
- Code 128
- Code 3 of 9
- 10 mil narrow bar element (x-dimension = 0.010")
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service