Skip to Content
Merck
All Photos(1)

Documents

CLS1500P250

Corning® narrow mouth reagent bottle, reusable

capacity 250 mL, polypropylene, with GL-45 PP screw cap

Synonym(s):

laboratory bottles, plastic bottles, reagent bottles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

cylindrical bottle
narrow-mouth bottle
polypropylene
screw top cap
translucent bottle

feature

cap

packaging

pack of 20 ea

manufacturer/tradename

Corning 1500P-250

H × O.D.

204 mm × 108 mm

capacity

250 mL

fitting

45

Looking for similar products? Visit Product Comparison Guide

General description

These bottles are manufactured from translucent polypropylene. They are supplied with PP screw top caps.

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service