CIB001
Cell-in-a-Box®
Cell Encapsulation Kit
Synonym(s):
Cell Culture Box
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
usage
(up to 5 experiments, each creating ~120 capsules, each containing between 4,000 and 40,000 cells.)
storage condition
room temperature
General description
The Cell-in-a-Box technology is a means to protect, isolate, store and transport living cells. This novel, manual encapsulation kit is a modified version of Cell-in-a-Box which acts as an introduction to this novel and previously unavailable proprietary technology. This kit allows one to encapsulate cells using proprietary materials and methods that can be performed manually by any end user using a simple disposable system. The kit allows researchers to quickly and easily encapsulate their own cells for further laboratory studies such as testing for cell survival, viability, growth, and release of biomolecules.
The Cell-in-a-Box kit aims to replace the current methods of cell encapsulation which involve the use of large, expensive, and technologically challenging machinery. Often this is very rarely available to the average scientist in a research setting and far too expensive to purchase for initial experimentation. The unique encapsulation material also replaces commonly available encapsulation materials which have technological drawbacks. This kit version of Cell-in-a-Box represents a major leap forward in the cell encapsulation field that has remained largely stagnant for the past 30 years.
The components of The Cell-in-a-Box kit are non-toxic and biocompatible. The porous beads are approximately 2 mm in diameter and have a pore size which allows nutrients to enter and waste products and biomolecules as large as antibodies to leave the capsule unhindered yet cells are unable to pass in or out. The capsules entrap the cells securely within their polymer membranes, yet allow the cells to continue growing within the confined boundaries. Nutrients and secreted factors are able to diffuse and exchange through the membrane pores, which can be selectively tuned.
The durable encapsulation material is made of biocompatible cellulose-based material and can last for years in vitro and in vivo (although kit-made capsules are not designed for in vivo use) as well as contribute to longer cell survival periods. This kit can be tested with any kind of living cells. Encapsulated cells can be frozen, stored, and transported.
Since the proprietary cell packaging material used for the Cell-in-a-Box technology is derived from a naturally occurring material, it is thus harmless to the environment, biologically inert, and has been shown to be safe in humans.
The Cell-in-a-Box kit aims to replace the current methods of cell encapsulation which involve the use of large, expensive, and technologically challenging machinery. Often this is very rarely available to the average scientist in a research setting and far too expensive to purchase for initial experimentation. The unique encapsulation material also replaces commonly available encapsulation materials which have technological drawbacks. This kit version of Cell-in-a-Box represents a major leap forward in the cell encapsulation field that has remained largely stagnant for the past 30 years.
The components of The Cell-in-a-Box kit are non-toxic and biocompatible. The porous beads are approximately 2 mm in diameter and have a pore size which allows nutrients to enter and waste products and biomolecules as large as antibodies to leave the capsule unhindered yet cells are unable to pass in or out. The capsules entrap the cells securely within their polymer membranes, yet allow the cells to continue growing within the confined boundaries. Nutrients and secreted factors are able to diffuse and exchange through the membrane pores, which can be selectively tuned.
The durable encapsulation material is made of biocompatible cellulose-based material and can last for years in vitro and in vivo (although kit-made capsules are not designed for in vivo use) as well as contribute to longer cell survival periods. This kit can be tested with any kind of living cells. Encapsulated cells can be frozen, stored, and transported.
Since the proprietary cell packaging material used for the Cell-in-a-Box technology is derived from a naturally occurring material, it is thus harmless to the environment, biologically inert, and has been shown to be safe in humans.
Application
The proprietary technology may be used for a variety of applications including:
The possibilities are only limited by your imagination.
- In healthcare, it facilitates the safe implantation of cells, holding them isolated from the immune system and allowing long term production of therapeutic molecules in patients.
- In biotechnology, the Cell-in-a-Box technology protects cells in bioreactors and fermenters, while at the same time simplifying purification of bioproducts produced from the cells and reducing production costs.
- In agriculture, the Cell-in-a Box technology could help improve the survival and growth of crops, as well as to increase crop yield and to prevent spoilage.
- Enhancement of protein expression systems.
- Implantation of hybridomas in mice.
- And many more.....
The possibilities are only limited by your imagination.
Features and Benefits
- Encapsulate any type of living cell.
- Can protect, isolate, store, and transport living cells.
- Semi-permeable and semi-transparent membrane.
- Biocompatible and non-toxic.
- Cells continue to grow inside ~2 mm bead in a 3-D environment.
- Straighforward procedure takes approximately 30 minutes.
- Cells can live inside capsules for extended periods.
- Capsules protect cells from graph vs host.
Packaging
Each kit includes 5 luer-lock syringes, 5 sterile blunt-end G34 droplet needles, 5 sterile sharp-end G18.5 filling needles, 5 mL of Solution 1, and 50 mL of Solution 2. Volumes of the solutions have been optimized for the cell encapsulation protocol and are not available for sale separately.
Legal Information
(a)This product shall only be used for research purposes. Purchaser shall not use the product to manufacture commercial products and/or to sell services relating to or using the product.
(b) Sigma Aldrich does not grant to purchaser any license, express or implied, in violation of or infringing upon any third party rights. Purchaser is responsible for determining the existence of any such third party rights and to acquire any such rights, should they exist, before using the product.
(c) Purchaser acknowledges and agrees that the product is not intended for use in humans, and agrees not to conduct research with it in humans.
(b) Sigma Aldrich does not grant to purchaser any license, express or implied, in violation of or infringing upon any third party rights. Purchaser is responsible for determining the existence of any such third party rights and to acquire any such rights, should they exist, before using the product.
(c) Purchaser acknowledges and agrees that the product is not intended for use in humans, and agrees not to conduct research with it in humans.
Cell-in-a-Box is a registered trademark of Austrianova
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Advances in experimental medicine and biology, 670, 92-103 (2010-04-14)
Although cell encapsulation technologies were originally developed for the treatment of acquired and genetic diseases such as diabetes, they can also be applied to the treatment of a variety of solid tumours. There are a number of strategies aimed at
Xenotransplantation, 13(4), 337-344 (2006-06-14)
Late diabetic complications cannot be prevented totally by current antidiabetic strategies. Therefore, new therapeutic concepts of insulin replacement such as pancreas transplantation are evolving. Due to the shortage of human donor organs, transplantation of microencapsulated xenogeneic pancreatic islet cells has
Current opinion in molecular therapeutics, 12(4), 450-460 (2010-08-03)
One of the first strategies for cancer gene therapy was the use of suicide gene/prodrug combinations, originally delivered to tumor cells using viral vectors. A major limitation of this approach was the inefficiency of suicide gene delivery. An alternative strategy
Lancet (London, England), 357(9268), 1591-1592 (2001-05-30)
Pancreatic cancer can seldom be resected, and chemotherapy has only a limited effect on survival or tumour load. We did a phase I/II trial in 14 patients with pancreatic cancer to assess the safety of local activation of low-dose ifosfamide.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service