Skip to Content
Merck
All Photos(1)

Key Documents

RT4-D6P2T

93011415, rat nerve, Bipolar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

RT4-D6P2T, 93011415

biological source

rat nerve

growth mode

Adherent

karyotype

Not specified

morphology

Bipolar

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat Schwann cell, immature

Cell Line Description

Derived from the ethylnitrosourea-induced tumour line D6 of the rat peripheral nervous system. Cells will de-differentiate after 10 passages. Read references prior to use.

Application

Schwann cell gene regulation and development

Culture Medium

DMEM + 2mM Glutamine + 5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service