Skip to Content
Merck
All Photos(1)

Key Documents

HSDM1C1

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

mouse fibrosarcoma

packaging

tube of 5 μg 90112810-DNA-5UG
pkg of vial of cells 90112810-1VL

growth mode

Adherent

karyotype

2n = 40

morphology

Fibroblast

products

Prostaglandin E2

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Swiss Albino fibrosarcoma

Cell Line Description

A clone derived from the parental HSDM1 cell line established from a transplantable fibrosarcoma of a Swiss Albino mouse. The cells secrete prostaglandin E2 which is inhibited by aspirin like drugs such as indomethacin.

Application

Hormone studies

Culture Medium

Ham′s F10 + 2mM Glutamine + 15% Horse Serum (HS) + 2.5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:4 i.e. seeding at 3-4x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service