Skip to Content
Merck
All Photos(1)

Documents

ML-1

88113007, human blood, Lymphoblastoid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

ML-1, 88113007

biological source

human blood

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblastoid

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human acute myeloblastic leukaemia

Cell Line Description

ML-1 is one of three celll lines isolated in 1978 from the peripheral blood of a 24 year old male patient with acute myeloblastic leukaemia. The cells can convert to more mature cells by the use of DMSO. The Y chromosome could not be detected in this cell line by short tandem repeat (STR)-PCR analysis when tested at ECACC. It is a known phenomenon that due to the increased genetic instability of cancer cell lines the Y chromosome can be rearranged or lost resulting in lack of detection. The cell line is identical to the source provided by the depositor based on the STR-PCR analysis.

Application

ML-1 has been used to compare the cell response to mafosfamide cyclohexylamine salt, 4-hydro-peroxy-cyclophosphamide and β-D-glucose-isophosphoramide mustard, which are oxazaphosphorine agents.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10,11
D13S317: 9,12
D16S539: 9,12
D5S818: 12
D7S820: 9,11
THO1: 7,9.3
TPOX: 8,10
vWA: 16

Subculture Routine

Maintain cultures between 2-9 x100,000 cells/ml; 5% CO2; 37°C. Freeze in 10% DMSO + 90% FBS. Immediately after resuscitation, pellet cells by centrifugation at 150 x g for 5 minutes and resuspend the cell pellet in fresh medium. This is to remove the presence of DMSO which may cause differentiation of the cells if allowed to remain.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service