Skip to Content
Merck
All Photos(1)

Documents

T47D/TR-1

16022513, human breast, polygonal epithelia

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

T47D/TR-1, 16022513, from human breast

biological source

human breast

growth mode

Adherent

morphology

polygonal epithelia

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

T47D/TR-1 cells are ER alpha positive and express progesterone receptor, although at reduced level compared to parental T7D/S2 cells. T47D/TR-1 cells are growth inhibited by fulvestrant. Oestrogen receptor.

Cell Line Description

The T47D/TR-1 is a breast cancer cell line resistant to tamoxifen. The T47D/TR-1 cell line has been established by long term treatment of T47D/S2 cells with 1 μM tamoxifen. Clonal selection was performed in medium without tamoxifen. After growth in presence of 1 μM tamoxifen for 10 months, the growth rate was similar to the parental cells.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4 x 10, 000 cells/cm² using 0.05% trypsin/EDTA; 5% CO2; 37°C. Recommend media changes every 2-3 days.

Other Notes

Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Citation

If use of this culture results in a scientific publication, it should be cited in the publication as: T47D/TR-1 (ECACC 16022513)

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service