Skip to Content
Merck
All Photos(1)

Key Documents

C5243

Sigma-Aldrich

Monoclonal Anti-Cystatin B antibody produced in mouse

~2 mg/mL, clone RJMW-2E7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CSTB, Anti-EPM1, Anti-PME, Anti-STEFIN B, Anti-STFB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RJMW-2E7, monoclonal

form

buffered aqueous solution

mol wt

antigen ~12 kDa

species reactivity

human, monkey

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1-2 μg/mL using extracts of MCF7 cells (is recommended using extracts of MCF7 cells.)

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CSTB(1476)

Immunogen

human cystatin B purified from human muscle and spleen.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chan Chung et al.
Human molecular genetics, 25(7), 1434-1446 (2016-02-26)
Niemann-Pick C1 (NPC) disease, an autosomal recessive lipid trafficking disorder caused by loss-of-function mutations in the NPC1 gene, is characterized by progressive neurodegeneration resulting in cognitive impairment, ataxia and early death. Little is known about the cellular pathways leading to
Kirsi Alakurtti et al.
European journal of human genetics : EJHG, 13(2), 208-215 (2004-10-16)
Loss-of-function mutations in the cystatin B (CSTB), a cysteine protease inhibitor, gene underlie progressive myoclonus epilepsy of Unverricht-Lundborg type (EPM1), characterized by myoclonic and tonic-clonic seizures, ataxia and a progressive course. A minisatellite repeat expansion in the promoter region of
A Azimi et al.
British journal of cancer, 110(10), 2489-2495 (2014-04-12)
Disseminated cutaneous malignant melanoma (CMM) is commonly unresponsive to standard chemotherapies, and there are as yet no predictive markers of therapy response. In the present study we collected fresh-frozen pretreatment lymph-node metastasis samples (n=14) from melanoma patients with differential response

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service