AXYMCT200R
Boil proof microtubes
size 2.0 mL, red, pkg of 10x500tubes/cs
Synonym(s):
centrifuge tubes
About This Item
Recommended Products
manufacturer/tradename
Axygen Inc MCT-200-R
Corning
packaging
pkg of 10x500tubes/cs
size
2.0 mL
color
red
Looking for similar products? Visit Product Comparison Guide
Related Categories
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service