Skip to Content
Merck
All Photos(1)

Key Documents

AV45164

Sigma-Aldrich

Anti-BTN1A1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-BT, Anti-BTN, Anti-Butyrophilin, subfamily 1, member A1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

56 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BTN1A1(696)

General description

Butyrophilin, subfamily 1, member A1, a BTN family member, is found in mammary tissue, spleen and thymus. Butyrophilin 1a1 (Btn1a1) is highly expressed in the lactating mammary gland and is secreted into milk in association with lipid droplets. Btn1a1 functions either as a structural protein or as a signaling receptor by binding to xanthine dehydrogenase/oxidase.

Specificity

Anti-Butyrophilin-1A1 polyclonal antibody reacts with human Butyrophilin-1A1 proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of human BTN1A1

Application

Anti-Butyrophilin-1A1 polyclonal antibody is used to tag Btn1a1 proteins for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques. It is used as a probe to study the role of Btn1a1 in lactation.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

Butyrophilin is the major protein associated with fat droplets in the milk. It is a member of the immunoglobulin superfamily. It may have a cell surface receptor function. The human butyrophilin gene is localized in the major histocompatibility complex (MHC) class I region of 6p and may have arisen relatively recently in evolution by the shuffling of exons between 2 ancestral gene families.Butyrophilin is the major protein associated with fat droplets in the milk. It is a member of the immunoglobulin superfamily. It may have a cell surface receptor function. The human butyrophilin gene is localized in the major histocompatibility complex (MHC) class I region of 6p and may have arisen relatively recently in evolution by the shuffling of exons between 2 ancestral gene families.

Sequence

Synthetic peptide located within the following region: LPCRLSPNASAEHLELRWFRKKVSPAVLVHRDGREQEAEQMPEYRGRATL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service