A8955
(S)-(+)-2-Amino-5-(iodoacetamido)pentanoic acid
≥90%
Synonym(s):
Nδ-Iodoacetyl-L-ornithine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C7H13N2O3I
CAS Number:
Molecular Weight:
300.09
MDL number:
UNSPSC Code:
12352200
Recommended Products
Assay
≥90%
storage temp.
−20°C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
C A Robertson et al.
Biochemical and biophysical research communications, 197(2), 523-528 (1993-12-15)
Nitric oxide synthase (EC 1.14.23) substrate analog inhibitors NG-monomethyl-L-Arg, NG-nitro-L-Arg, NG-nitro-L-Arg methyl ester, and aminoguanidine were examined as potential inhibitors of rat liver arginase (EC 3.5.3.1). NG-nitro-L-Arg was found to inhibit arginase catalyzed conversion of L-Arg to L-Orn at pH
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service