Skip to Content
Merck
All Photos(1)

Key Documents

A4854

Sigma-Aldrich

Anti-β3 Adrenergic Receptor antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 17 μg/mL using human adipose tissue

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADRB3(155)
mouse ... Adrb3(11556)
rat ... Adrb3(25645)

General description

Adrenergic receptors are a G-protien coupled receptors that exist in two forms, alpha or beta. Ligands for adrenergic receptors are catecholamines that induce a sympathetic response in animal cells.
Rabbit Anti-β3 Adrenergic Receptor specifically recognizes human β3 adrenergic receptor.

Immunogen

synthetic peptide corresponding to N-terminal extracellular domain of human β3 adrenergic receptor. The immunizing peptide has 80% homology with the rat gene and 75% homology with the mouse gene.

Application

Rabbit Anti-β3 Adrenergic Receptor can be used (at 17μg/mL) for immunohistochemistry assays using human adipose tissue.

Physical form

solution in phosphate buffered saline, pH 7.7, containing 0.1% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrés Pun-García et al.
Basic research in cardiology, 117(1), 62-62 (2022-11-30)
Aortic stenosis (AS) is associated with left ventricular (LV) hypertrophy and heart failure (HF). There is a lack of therapies able to prevent/revert AS-induced HF. Beta3 adrenergic receptor (β3AR) signaling is beneficial in several forms of HF. Here, we studied

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service