Skip to Content
Merck
All Photos(1)

Key Documents

A2164

Sigma-Aldrich

Albumin, Sulforhodamine 101 acid chloride bovine

lyophilized powder

Synonym(s):

Texas Red-Albumin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202

biological source

bovine

form

lyophilized powder

extent of labeling

2-3 mol sulforhodamine 101 acid chloride per mol albumin

impurities

salt, free

storage temp.

2-8°C

Application

Albumin conjugated to Texas Red was used to evaluate microvascular permeability in a human tumor xenograft, but it is a general method for use in human and animal superficial tumors.

Preparation Note

Prepared from crystallized and lyophilized albumin (A4378).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service