Skip to Content
Merck
All Photos(1)

Documents

A0894

Sigma-Aldrich

5′-Amino-2′,5′-dideoxy-5-iodouridine

~96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12IN3O4
CAS Number:
Molecular Weight:
353.11
UNSPSC Code:
12352204

Assay

~96%

storage temp.

2-8°C

Biochem/physiol Actions

Potent inhibitor of herpes simplex virus (HSV) in cell culture.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y C Cheng et al.
Journal of virology, 15(5), 1284-1285 (1975-05-01)
5-Amino-2,5-dideoxy-5-iodouridine, a nel thymidine analogue, is a potent inhibitor of herpes simplex virus type 1 replication. In contrast to most other nucleoside analogues which possess antiviral activity, 5-amino-2,5-dideoxy-5-iodouridine exhibits little, if any, cellular toxicity. Preliminary evidence suggests that 5-amino-2,5-dideoxy-5-iodouridine selectively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service