80556
Calcium chloride 0.19 M – HEPES-Na pH 7.5; 0.095M – Polyethylene glycol 400; 26.6% (w/v) – Glycerol 5% solution
Synonym(s):
Crystallization solution 14/Kit No 75403
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
form
liquid
pH
7.5±0.2 ( in neat)
density
1.09 g/mL at 20 °C
storage temp.
2-8°C
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service