Skip to Content
Merck
All Photos(1)

Key Documents

73342

Sigma-Aldrich

β-Carotene 5,6-epoxide

≥85.0% (HPLC)

Synonym(s):

(5RS,6RS)-5,6-Epoxy-5,6-dihydro-β,β-carotene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H56O
CAS Number:
Molecular Weight:
552.87
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Assay

≥85.0% (HPLC)

form

powder

λ

in hexane (with 2% dichloromethane)

UV absorption

λ: 443-447 nm Amax

storage temp.

−20°C

SMILES string

CC(/C=C/C1=C(C)CCCC1(C)C)=C\C=C\C(C)=C\C=C\C=C(C)\C=C\C=C(\C=C\C23C(C)(C)CCCC2(O3)C)C

InChI

1S/C40H56O/c1-31(19-13-21-33(3)24-25-36-35(5)23-15-27-37(36,6)7)17-11-12-18-32(2)20-14-22-34(4)26-30-40-38(8,9)28-16-29-39(40,10)41-40/h11-14,17-22,24-26,30H,15-16,23,27-29H2,1-10H3/b12-11+,19-13+,20-14+,25-24+,30-26+,31-17+,32-18+,33-21+,34-22+

InChI key

RVCRIPILOFSMFG-WWSVUWEKSA-N

General description

The 5,6-epoxy structure can give rise to a wide variety of carotenoid end-groups with the plausible biosynthetic transformations of the 5,6-epoxy-ß-ring to give diverse structures such as the cyclopentane carotenoids capsanthin and capsorubin, the retro-carotenoids eschscholtz- xanthin and rhodoxanthin, allenic carotenoids like the neoxanthin and fucoxanthin, 5,6-diols like the karpoxanthin and latoxanthin, 3,6-poxides as found in eutreptiellanone and cucurbitaxanthin, the 6-hydroxy-γ-ring structure found in prasinoxanthin and nigroxanthin, as well as the 5,6-seco-5,6-dione-end-group of semi-ß-carotenone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandra Gentili et al.
Journal of agricultural and food chemistry, 63(33), 7428-7439 (2015-08-11)
This paper presents an analytical strategy for a large-scale screening of carotenoids in tomato fruits by exploiting the potentialities of the triple quadrupole-linear ion trap hybrid mass spectrometer (QqQLIT). The method involves separation on C30 reversed-phase column and identification by
G Martano et al.
Analytical and bioanalytical chemistry, 406(12), 2909-2924 (2014-03-22)
A validated ultrahigh-performance liquid chromatography method using 1.7 μm core-shell particles is presented for the identification and quantification of β-carotene (BC) and related cleavage products (CPs) in primary cell culture media. Besides BC, apo-4'-, apo-8'-, apo-10'-, and apo-12'-carotenals, as well
Poliana D Gurak et al.
Food chemistry, 147, 160-169 (2013-11-12)
In this study HPLC-DAD-MS/MS was applied for the identification of compounds derived from (all-E)-β-carotene following epoxidation and oxidative cleavage. The consequences on the CIELAB colour parameters and antioxidant capacity (AC) were also evaluated. Five apocarotenoids, three secocarotenoids, seven Z isomers
T A Kennedy et al.
The Journal of biological chemistry, 267(7), 4658-4663 (1992-03-05)
The effect of the partial pressure of oxygen (pO2) on the antioxidant reactions of all-trans-beta, beta-carotene (BC) was investigated in a soybean phosphatidylcholine liposome system. Peroxyl radicals generated by thermolysis of azo-bis(2,4-dimethylvaleronitrile) at 37 degrees C initiated lipid peroxidation. BC
Oxidative degradation of ?-carotene and ?-apo-8'-carotenal
Mordi, R.C., et al.
Tetrahedron, 49, 911-928 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service