55461
O-(Decylphosphoryl)choline solution
Synonym(s):
Phosphocholine decyl ester
About This Item
description
zwitterionic
mol wt
323.41 g/mol
concentration
100 mM in H2O
technique(s)
protein quantification: suitable
application(s)
detection
storage temp.
2-8°C
SMILES string
[O-]P(OCC[N+](C)(C)C)(OCCCCCCCCCC)=O
InChI
1S/C15H34NO4P/c1-5-6-7-8-9-10-11-12-14-19-21(17,18)20-15-13-16(2,3)4/h5-15H2,1-4H3
InChI key
VVVDGSCGBGBGFN-UHFFFAOYSA-N
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service