Skip to Content
Merck
All Photos(1)

Key Documents

31005

Sigma-Aldrich

Mercury(II) chloride

puriss. p.a., ACS reagent, reag. ISO, ≥99.5% (calc. to the dried substance)

Synonym(s):

Mercuric chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HgCl2
CAS Number:
Molecular Weight:
271.50
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥99.5% (calc. to the dried substance)
vapor pressure:
1.3 mmHg ( 236 °C)

grade

ACS reagent
puriss. p.a.

Agency

USP/NF
reag. ISO

vapor pressure

1.3 mmHg ( 236 °C)

Assay

≥99.5% (calc. to the dried substance)

impurities

≤0.001% other heavy metals (as Pb)
≤0.002% total nitrogen (N)
≤0.05% mercury(I)-chloride

ign. residue (after red. with HCOOH)

≤0.01% (as SO4)

loss

≤1% loss on drying (on P2O5)

mp

277 °C (lit.)

anion traces

sulfate (SO42-): ≤200 mg/kg

cation traces

Ag: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Cu: ≤10 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤10 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤10 mg/kg
Zn: ≤5 mg/kg

SMILES string

Cl[Hg]Cl

InChI

1S/2ClH.Hg/h2*1H;/q;;+2/p-2

InChI key

LWJROJCJINYWOX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Muta. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Combined use of ISCR and biostimulation techniques in incomplete processes of reductive dehalogenation of chlorinated solvents.
Jofre Herrero et al.
The Science of the total environment, 648, 819-829 (2018-08-24)
Alexey Vorobev et al.
Applied and environmental microbiology, 79(19), 5918-5926 (2013-07-23)
Many methanotrophs have been shown to synthesize methanobactin, a novel biogenic copper-chelating agent or chalkophore. Methanobactin binds copper via two heterocyclic rings with associated enethiol groups. The structure of methanobactin suggests that it can bind other metals, including mercury. Here
Masaro Akai et al.
Journal of bacteriology, 194(24), 6828-6836 (2012-10-09)
The moderately halotolerant cyanobacterium Synechocystis sp. strain PCC 6803 contains a plasma membrane aquaporin, AqpZ. We previously reported that AqpZ plays a role in glucose metabolism under photomixotrophic growth conditions, suggesting involvement of AqpZ in cytosolic osmolarity homeostasis. To further
Minghua Wang et al.
Aquatic toxicology (Amsterdam, Netherlands), 130-131, 123-131 (2013-02-19)
Mercury (Hg) is a widespread persistent pollutant in aquatic ecosystems. We investigated the protein profiles of medaka (Oryzias melastigma) liver chronically exposed to different mercuric chloride (HgCl2) concentrations (1 or 10 μg/L) for 60 d using two-dimensional difference gel electrophoresis
Fenglian Xu et al.
Molecular brain, 5, 30-30 (2012-09-18)
Mercury is a well-known neurotoxin implicated in a wide range of neurological or psychiatric disorders including autism spectrum disorders, Alzheimer's disease, Parkinson's disease, epilepsy, depression, mood disorders and tremor. Mercury-induced neuronal degeneration is thought to invoke glutamate-mediated excitotoxicity, however, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service