Skip to Content
Merck
All Photos(1)

Documents

PHR1959

Supelco

Salbutamol Impurity B

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(1RS)-2-[(1,1-Dimethylethyl)amino]-1-(4-hydroxyphenyl)ethanol, Albuterol Related Compound I, 2-tert-Butylamino-1-(4-hydroxyphenyl)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19NO2
CAS Number:
Molecular Weight:
209.28
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 852
traceable to Ph. Eur. Y0000030

form

powder

CofA

current certificate can be downloaded

packaging

pkg of 50 mg

application(s)

pharmaceutical

storage temp.

2-8°C

InChI

1S/C12H19NO2/c1-12(2,3)13-8-11(15)9-4-6-10(14)7-5-9/h4-7,11,13-15H,8H2,1-3H3

InChI key

JOGFUYPGDLRKHD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application


  • Salbutamol Impurity B Analysis in Pharmaceutical Products: Utilizing advanced chromatographic techniques, researchers have developed validated methods for the precise identification and quantification of Salbutamol Impurity B in pharmaceutical formulations. This critical quality control measure ensures the safety and efficacy of bronchodilator medications, adhering to stringent regulatory standards (Kondra et al., 2023).

  • Development of Analytical Standards for Salbutamol Impurity B: The quantification of Salbutamol Impurity B using achiral supercritical fluid chromatography represents a significant advancement in the field of pharmaceutical impurities identification. This method provides a robust, efficient, and reproducible approach for monitoring the stability and purity of salbutamol-based therapies, contributing to enhanced patient safety (Dispas et al., 2017).

  • Research on Salbutamol Degradation Products: A stability-indicating RP-HPLC method has been developed for the simultaneous determination of Salbutamol Impurity B and other related substances in nasal solutions. This method is crucial for understanding the degradation pathways of salbutamol, thereby assisting in the development of more stable and effective bronchodilator formulations (Kasawar & Farooqui, 2010).

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB3289 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service