Skip to Content
Merck
All Photos(1)

Documents

NISTRM8557

NBS 127 (sulfur and oxygen isotopes in barium sulfate)

NIST® RM 8557

Synonym(s):

Barium sulfate, Baryte

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaSO4
CAS Number:
Molecular Weight:
233.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

isotopic purity

 18O
 34S

manufacturer/tradename

NIST®

format

matrix material

SMILES string

[Ba++].[O-]S([O-])(=O)=O

InChI

1S/Ba.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

TZCXTZWJZNENPQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Analyte : Sulfur (34S), Oxygen (18O)

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service