Skip to Content
Merck
All Photos(1)

Documents

NIST967A

Creatinine in frozen human serum

NIST® SRM® 967a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

packaging

pkg of set(4) (2 ea conc)

manufacturer/tradename

NIST®

technique(s)

mass spectrometry (MS): suitable

application(s)

clinical testing

format

matrix material

General description

This human blood serum creatinine is a standard reference material (SRM) that consists of four stoppered vials of frozen human serum, two vials each at two different creatinine concentration levels. Each vial contains 1.0 mL of human serum.

SRM 967A_cert

SRM 967A_SDS

Application

This human blood serum creatinine is intended primarily for use in evaluating the accuracy of procedures for the determination of creatinine in human serum. It is also meant for use in validating working or secondary reference materials.

Other Notes

  • For more information on storage, usage, and expiry refer to the NIST certificate.
  • Notes for biomaterials, disposal, and transport information are available in SDS.
Certified for the analytes listed below.
Creatinine

See certificate for values and more details at nist.gov/SRM.

Preparation Note

  • The serum is shipped frozen on dry ice and, upon receipt, should be stored frozen until ready for use.
  • Vials of the SRM to be analyzed should be removed from the freezer and allowed to stand at room temperature (20 °C to 25 °C) until thawed.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Greg Miller et al.
Advances in chronic kidney disease, 25(1), 7-13 (2018-03-04)
In 2002, the Kidney Disease Outcomes Quality Initiative guidelines for identifying and treating CKD recommended that clinical laboratories report estimated glomerular filtration rate (eGFR) with every creatinine result to assist clinical practitioners to identify people with early-stage CKD. At that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service