Skip to Content
Merck
All Photos(1)

Documents

NIST8403

Cocoa Flavanol Extract

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

packaging

pkg of 10 g

manufacturer/tradename

NIST®

application(s)

food and beverages

format

matrix material

storage temp.

-10 to -25°C

General description

RM 8403 is a free-flowing powder of cocoa flavanols, and it is sealed in an aluminized mylar sachets.

Each unit of RM 8403 consists of five sachets each containing approximately 2 g of the material.

SRM 8403 _SDS

SRM 8403 _CoA

Application

Cocoa Flavanol Extract reference material (RM) is intended for harmonizing methods of the determination of cocoa flavanols and their oligomers, including those with a degree of polymerization (DP) of up to seven units.

Features and Benefits

  • RM 8403 is available with non-certified mass fraction values of degree of polymerization of Cocoa flavanol extract.
  • Non-certified values are measured using liquid chromatography with fluorescence detection. 
  • The NIST certificate is provided with safety, storage, and use instructions.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service