ERMBF417B
Maize GMO Standard MON 863 x MON 810
for the detection of GMO Maize (MON 863 × MON 810) with PCR
Synonym(s):
GMO Maize Powder Standard 0% MON 863 x MON 810
About This Item
Recommended Products
grade
certified reference material
product line
BioChemika
quality
for the detection of GMO Maize (MON 863 × MON 810) with PCR
composition
genetically modified MON 863 × MON 810 maize, 0.1%
manufacturer/tradename
JRC
application(s)
cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)
format
matrix material
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Analysis Note
ERMBF417B
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service