Skip to Content
Merck
All Photos(1)

Key Documents

94901

Sigma-Aldrich

9-Vinylanthracene

technical, ≥95.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12
CAS Number:
Molecular Weight:
204.27
Beilstein:
2041641
EC Number:
MDL number:
UNSPSC Code:
12162002

grade

technical

Assay

≥95.0% (GC)

bp

61-66 °C/10 mmHg (lit.)

mp

62-65 °C (lit.)
64-66 °C

storage temp.

2-8°C

SMILES string

C=Cc1c2ccccc2cc3ccccc13

InChI

1S/C16H12/c1-2-14-15-9-5-3-7-12(15)11-13-8-4-6-10-16(13)14/h2-11H,1H2

InChI key

OGOYZCQQQFAGRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service