Skip to Content
Merck
All Photos(1)

Documents

94423

Millipore

Legionella Enrichment Broth Base

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

liver extract, 2.5 g/L
proteose peptone, 15 g/L
sodium chloride, 5 g/L
yeast extract, 5 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

6.9±0.2 (25 °C)

application(s)

environmental
water monitoring

microbiology

suitability

Legionella spp.

Related Categories

Application

Legionella Enrichment Broth Base with addition of supplements is used for the enrichment of Legionella species

Preparation Note

Suspend 13.75 grams in 500 ml distilled water. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. Do not heat prior to sterilization. Cool to 50°C and aseptically add 1 vial of Legionella Growth Supplement Part A and Part B (Twin Pack, Cat. No. 42981).

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service