Skip to Content
Merck
All Photos(1)

Key Documents

93438

Sigma-Aldrich

Tris(trimethylsilyl)amine

purum, ≥97.0% (GC)

Synonym(s):

Nitrilotris(trimethylsilane)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3Si]3N
CAS Number:
Molecular Weight:
233.57
Beilstein:
1851880
EC Number:
MDL number:
UNSPSC Code:
12352103

grade

purum

Assay

≥97.0% (GC)

bp

72 °C/10 mmHg (lit.)

mp

65-69 °C
67-69 °C (lit.)

SMILES string

C[Si](C)(C)N([Si](C)(C)C)[Si](C)(C)C

InChI

1S/C9H27NSi3/c1-11(2,3)10(12(4,5)6)13(7,8)9/h1-9H3

InChI key

PEGHITPVRNZWSI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the preparation of N-trimethylsilylated nitrido-metal and nitrido-non-metal complexes from halides

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Rhiel et al.
Angewandte Chemie (International Edition in English), 106, 599-599 (1994)
H.W. Roesky et al.
Zeitschrift fur Naturforschung, B: Chemical Sciences, 43, 72-72 (1988)
K. Niedenzu et al.
Zeitschrift fur Naturforschung, B: Chemical Sciences, 47, 713-713 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service