Skip to Content
Merck
All Photos(1)

Key Documents

92824

Sigma-Aldrich

Picrylsulfonic acid solution

purum, ~1% in DMF

Synonym(s):

2,4,6-Trinitrobenzenesulfonic acid solution, TNBS

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H3N3O9S
CAS Number:
Molecular Weight:
293.17
Beilstein:
572358
MDL number:
UNSPSC Code:
12352200

grade

purum

concentration

~1% in DMF

refractive index

n20/D 1.431

storage temp.

2-8°C

SMILES string

OS(=O)(=O)c1c(cc(cc1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H3N3O9S/c10-7(11)3-1-4(8(12)13)6(19(16,17)18)5(2-3)9(14)15/h1-2H,(H,16,17,18)

InChI key

NHJVRSWLHSJWIN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hydrophilic modifying reagent. Derivatives with amino compounds can be regenerated with hydrazine.

Other Notes

Reagent used in a micro-test for detecting incomplete coupling in solid-phase peptide synthesis

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new micro-test for the detection of incomplete coupling reactions in solid-phase peptide synthesis using 2,4,6-trinitrobenzenesulphonic acid.
W S Hancock et al.
Analytical biochemistry, 71(1), 260-264 (1976-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service