Skip to Content
Merck
All Photos(1)

Key Documents

91541

SAFC

Oleic acid

Pharma Manufacturing

Synonym(s):

cis-9-Octadecenoic acid, Elainic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOH
CAS Number:
Molecular Weight:
282.46
Beilstein:
1726542
EC Number:
MDL number:
UNSPSC Code:
12352211
Pricing and availability is not currently available.

vapor pressure

1 mmHg ( 176 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

concentration

≥98 % (w/w) (GC)

technique(s)

cell culture | mammalian: suitable

impurities

≤0.4% water

ign. residue

≤0.01%

refractive index

n20/D 1.459 (lit.)

bp

194-195 °C/1.2 mmHg (lit.)

mp

13-14 °C (lit.)

acid value

190‑205 KOH/g

iodine value

85‑105

peroxide value

≤10.0

transition temp

freezing point 10-16 °C

density

0.889-0.895 g/mL at 20 °C
0.89 g/mL at 25 °C (lit.)

cation traces

As: ≤2 mg/kg
heavy metals: ≤20 mg/kg

storage temp.

2-8°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(O)=O

InChI

1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H,19,20)/b10-9-

InChI key

ZQPPMHVWECSIRJ-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

To request documentation for this product, please contact Customer Support and select ‘Product Documentation′. Please note that access to documentation for this product requires a confidentiality disclosure agreement.

Analysis Note

remarks on GC: content of fatty acid pass
pharmacopoea tests: mineral acid pass

microbiol contamination ≤ 1000 CFU/g
residual solvents (GLC-HS) conforms to ICH Q3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacob F Smith et al.
Frontiers in physiology, 11, 80-80 (2020-03-07)
Osteoarthritis (OA) is a degenerative joint disease characterized by inflammatory degradation of articular cartilage and subchondral bone. Wogonin, a compound extracted from the plant Scutellaria baicalensis (colloquially known as skullcap), has previously been shown to have direct anti-inflammatory and antioxidative

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service