Skip to Content
Merck
All Photos(1)

Key Documents

909513

Sigma-Aldrich

3-Azido-7-hydroxycoumarin

≥95%

Synonym(s):

3-Azido-7-hydroxy-chroman-2-one, 3-Azido-7-hydroxy-chromen-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5N3O3
CAS Number:
Molecular Weight:
203.15
MDL number:
UNSPSC Code:
12352202
form:
solid
Assay:
≥95%
Pricing and availability is not currently available.

Assay

≥95%

form

solid

availability

available only in USA

storage temp.

−20°C

InChI

1S/C9H5N3O3/c10-12-11-7-3-5-1-2-6(13)4-8(5)15-9(7)14/h1-4,13H

InChI key

KKJNQKVCZCNJDI-UHFFFAOYSA-N

Application

This special UV active chromophore is ideal for imaging purposes after conjugation to biomolecules. It emits fluorescence only after the click reaction when its electron system is expanded. Therefore no background signal is expected for the unreacted compound. λabs 404 nm (after click reaction), λem 477 nm (after click reaction ).

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott T Laughlin et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(1), 12-17 (2008-12-24)
Molecular imaging enables visualization of specific molecules in vivo and without substantial perturbation to the target molecule's environment. Glycans are appealing targets for molecular imaging but are inaccessible with conventional approaches. Classic methods for monitoring glycans rely on molecular recognition

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service