Skip to Content
Merck
All Photos(1)

Key Documents

90850

Sigma-Aldrich

2,4,6-Tri-tert-butylpyridine

purum, ≥98.0% (CHN)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H29N
CAS Number:
Molecular Weight:
247.42
Beilstein:
1620027
MDL number:
UNSPSC Code:
12352100

grade

purum

Assay

≥98.0% (CHN)

bp

115-120 °C/20 mmHg (lit.)

mp

67-71 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)c1cc(nc(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C17H29N/c1-15(2,3)12-10-13(16(4,5)6)18-14(11-12)17(7,8)9/h10-11H,1-9H3

InChI key

IDOFMGSESVXXQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Strongly hindered base (proton trap); It undergoes nearly no alkylation

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y.V. Voznyi et al.
Carbohydrate Research, 54, 300-300 (1977)
H. Weber et al.
Chemische Berichte, 114, 1455-1455 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service