Skip to Content
Merck
All Photos(1)

Documents

90430

Sigma-Aldrich

Triethylene glycol di(p-toluenesulfonate)

purum, ≥98.0% (HPLC)

Synonym(s):

2,2′-(Ethylenedioxy)diethyl ditosylate, Triethylene glycol ditosylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3C6H4SO3CH2CH2OCH2-)2
CAS Number:
Molecular Weight:
458.55
Beilstein:
632509
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥98.0% (HPLC)

mp

78-82 °C (lit.)
78-82 °C

SMILES string

Cc1ccc(cc1)S(=O)(=O)OCCOCCOCCOS(=O)(=O)c2ccc(C)cc2

InChI

1S/C20H26O8S2/c1-17-3-7-19(8-4-17)29(21,22)27-15-13-25-11-12-26-14-16-28-30(23,24)20-9-5-18(2)6-10-20/h3-10H,11-16H2,1-2H3

InChI key

KCONMNWPRXAWKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Versatile building block for the synthesis of crown ethers, cryptands, and analogues

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G.W. Gokel et al.
Macrocyclic Polyether Synthesis (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service