901784
Nitro-grela 1st generation
≥95%
Synonym(s):
Dichloro(2-isopropoxy-5-nitrophenylmethylene)(tricyclohexylphosphine)ruthenium(II)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
≥95%
form
powder
reaction suitability
core: ruthenium
reagent type: catalyst
availability
available only in USA
mp
>300 °C
storage temp.
2-8°C
Application
Nitro-grela 1st generation is good for olefin metathesis for en-yn reactions.
Legal Information
Sold in collaboration with Apeiron Synthesis, Inc. Research quantities are 100 g / customer / year. For quantities that exceed the research limit, please contact the supplier at www.apeiron-synthesis.com.
PCT/EP2003/011222
PCT/EP2003/011222
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Self-react. C
Storage Class Code
5.2 - Organic peroxides and self-reacting hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of the American Chemical Society, 126(30), 9318-9325 (2004-07-30)
The design, synthesis, stability, and catalytic activity of nitro-substituted Hoveyda-Grubbs metathesis catalysts are described. The highly active and stable meta- and para-substituted complexes are attractive from a practical point of view. These catalysts operate in very mild conditions and can
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service