Skip to Content
Merck
All Photos(1)

Key Documents

89789

Supelco

Dipropylammonium acetate solution

0.5 M in H2O, suitable for ion pair chromatography

Synonym(s):

Dipropylamine acetate salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2)2NH · CH3COOH
CAS Number:
Molecular Weight:
161.24
Beilstein:
7937734
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
Pricing and availability is not currently available.

description

concentrate

Quality Level

form

liquid

packaging

ampule of 25 mL

concentration

0.5 M in H2O

technique(s)

ion pair chromatography: suitable

SMILES string

CC(O)=O.CCCNCCC

InChI

1S/C6H15N.C2H4O2/c1-3-5-7-6-4-2;1-2(3)4/h7H,3-6H2,1-2H3;1H3,(H,3,4)

InChI key

LKDMDHMCNGRIAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

concentrate in ampules for 6x1 l 0.005 M ready-to-use solution

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service