Skip to Content
Merck
All Photos(1)

Documents

87578

Supelco

Tetrakis(decyl)ammonium bromide

suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)

Synonym(s):

Tetra-decylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)9]4N(Br)
CAS Number:
Molecular Weight:
659.01
Beilstein:
5842649
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21

description

cationic

Quality Level

Assay

≥99.0% (AT)

form

powder

quality

LiChropur

mol wt

659.01 g/mol

technique(s)

ion pair chromatography: suitable

mp

87-90 °C

λ

10 % in methanol

UV absorption

λ: 240 nm Amax: 0.04
λ: 250 nm Amax: 0.03
λ: 260 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for filter test

SMILES string

[Br-].CCCCCCCCCC[N+](CCCCCCCCCC)(CCCCCCCCCC)CCCCCCCCCC

InChI

1S/C40H84N.BrH/c1-5-9-13-17-21-25-29-33-37-41(38-34-30-26-22-18-14-10-6-2,39-35-31-27-23-19-15-11-7-3)40-36-32-28-24-20-16-12-8-4;/h5-40H2,1-4H3;1H/q+1;/p-1

InChI key

AHNISXOXSNAHBZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Ion-pairing reagents as mobile phase additives enable the separation of ionic and highly polar substances on reversed phase HPLC columns. The purity of mobile phase additives is a most important parameter to be considered for their successful application. Sigma-Aldrich has a broad-range of tailor-made reagents for anionic (quaternary ammonium and phosphonium salts) and cationic (alkanesulfonates) determinations. All mobile phase additives are subjected to rigorous testing with special emphasis on the needs of modern reversed phase HPLC:

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service