Skip to Content
Merck
All Photos(2)

Key Documents

78889

Supelco

Osthole

analytical standard

Synonym(s):

7-Methoxy-8-(3-methyl-2-butenyl)coumarin, 7-Methoxy-8-isopentenylcoumarin, Osthol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16O3
CAS Number:
Molecular Weight:
244.29
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc2C=CC(=O)Oc2c1C\C=C(\C)C

InChI

1S/C15H16O3/c1-10(2)4-7-12-13(17-3)8-5-11-6-9-14(16)18-15(11)12/h4-6,8-9H,7H2,1-3H3

InChI key

MBRLOUHOWLUMFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Active constituent of Cnidium monnieri, a traditional Chinese medicine used for its anti-inflammatory, analgesic, and anti-lipedemic activities. Dramatically decreased lipid accumulation in a quail model. Its neuroprotective effect on MPP(+)-induced cytotoxicity in PC12 cells supports the use of osthole as a therapeutic agent for the treatment of neurodegenerative disorders. Suppression of fatty acid synthase expression in HER2-overexpressing breast cancer cells indicates osthole as a promising agent for chemotherapy.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ola D Alabi et al.
Life sciences, 102(2), 105-110 (2014-03-25)
Osthole, a coumarin derivative, has been used in Chinese medicine and studies have suggested a potential use in treatment of diabetes and cancers. Therefore, we investigated the effects of osthole and other coumarins on GLUT1 activity in two cell lines

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service