Skip to Content
Merck
All Photos(1)

Key Documents

70150

Supelco

Nafion NR 50

H+-form

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000

loss

~2% loss on drying

particle size

7-9 mesh

Other Notes

Nafion is a super-acidic perfluorinated resin-sulfonic acid used as catalyst for Friedel-Crafts type acylations; Acetalization and de-acetalization reactions; O-Silylations with allylsilanes

Legal Information

Nafion is a trademark of The Chemours Company FC, LLC

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K.S. Petrakis et al.
Superacids, 891-891 (1983)
G.A. Olah
Superacids (1985)
G.A. Olah et al.
The Journal of Organic Chemistry, 48, 3359-3359 (1983)
G.A. Olah et al.
Synthesis, 892-892 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service