69440
7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene
purum, ≥98.0% (GC)
Synonym(s):
1,3,4,6,7,8-Hexahydro-1-methyl-2H-pyrimido[1,2-a]pyrimidine, MTBD
About This Item
Recommended Products
grade
purum
Assay
≥98.0% (GC)
refractive index
n20/D 1.537 (lit.)
n20/D 1.537
bp
75-79 °C/0.1 mmHg (lit.)
density
1.067 g/mL at 25 °C (lit.)
SMILES string
CN1CCCN2CCCN=C12
InChI
1S/C8H15N3/c1-10-5-3-7-11-6-2-4-9-8(10)11/h2-7H2,1H3
InChI key
OEBXWWBYZJNKRK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
replaced by
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service