Skip to Content
Merck
All Photos(1)

Key Documents

64280

Sigma-Aldrich

Methanesulfonic acid

≥99.0%, ReagentPlus®

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO3H
CAS Number:
Molecular Weight:
96.11
Beilstein:
1446024
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

vapor density

3.3 (vs air)

vapor pressure

1 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99.0%

analyte chemical class(es)

amino acids

refractive index

n20/D 1.429 (lit.)
n20/D 1.429

bp

167 °C/10 mmHg (lit.)

mp

17-19 °C (lit.)

density

1.481 g/mL at 25 °C (lit.)

λ

neat

UV absorption

λ: 240-320 nm Amax: <0.4

SMILES string

[S](=O)(=O)(O)c2c(cccc2)C=C.c1(c(cccc1)C=C)C=C
O=S(O)(C)=O

InChI

1S/CH4O3S/c1-5(2,3)4/h1H3,(H,2,3,4)

InChI key

AFVFQIVMOAPDHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

372.2 °F - closed cup

Flash Point(C)

189 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

  1. Which document(s) contains shelf-life or expiration date information for a given product?

    If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.

  2. How do I get lot-specific information or a Certificate of Analysis?

    The lot specific COA document can be found by entering the lot number above under the "Documents" section.

  3. How do I find price and availability?

    There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers.

  4. What is the Department of Transportation shipping information for this product?

    Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product. 

  5. I received Product 64280, Methanesulfonic acid, and it was a solid.

    If the material is received as a solid, product 64280 is not compromised.  The material can be warmed to 25-30°C to melt the solid.

  6. What is the concentration of Product 64280, Methanesulfonic acid?

    The product 64280 is not a solution, but the pure product.  To calculate the molarity of the material, one would use the density and molecular weight.  For methanesulfonic acid the molarity is ~15.4M.

  7. What is Product 64280, Methanesulfonic acid, packaged in?

    The product 64280 is going to be packaged in amber glass bottle for the 25mL, 100mL, and 500mL packaging size.

  8. What type of containers should I avoid for Product 64280, Methanesulfonic acid?

    The product 64280 is corrosive to iron, steel, brass, copper, and lead.

  9. What can I dissolve Product 64280, Methanesulfonic acid, into?

    Product 64280 can be dissolved in hexane, 26-28; benzene, 1.50; toluene, 0.38; o-chlorotoluene, 0.23; ethyl disulfide, 0.47; these values are in weight %.

  10. How should Product 64280, Methanesulfonic acid, be handled?

    We recommend to handle and store product 64280 in a dry inert atmosphere.  Please refer to the MSDS or Technical Bulletin AL-134 for additional handling information.

  11. My question is not addressed here, how can I contact Technical Service for assistance?

    Ask a Scientist here.

Formation of sulfur dioxide and methanesulfonic acid in the photooxidation of dimethyl sulfide in the air.
Hatakeyama S, et al.
Geophysical Research Letters, 9(5), 583-586 (1982)
Methanesulfonic acid solution as supporting electrolyte for zinc-vanadium redox battery.
Tang C and Zhou D.
Electrochimica Acta, 65, 179-184 (2012)
Environmental benefits of methanesulfonic acid. Comparative properties and advantages.
Gernon MD.
Green Chemistry, 1(3), 127-140 (1999)
Phosphorus pentoxide-methanesulfonic acid. Convenient alternative to polyphosphoric acid.
Eaton PE, et al.
The Journal of Organic Chemistry, 38(23), 4071-4073 (1973)
Chris Twelves et al.
Breast cancer research and treatment, 148(3), 553-561 (2014-11-09)
Data from two phase 3 studies of eribulin were pooled in analyses initially requested by the European Medicines Agency to assess whether specific patient subgroups, previously treated with an anthracycline and a taxane, benefited from eribulin. Study 305/EMBRACE included women

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service