Skip to Content
Merck
All Photos(1)

Key Documents

62491

Sigma-Aldrich

Lithium diisopropylamide solution

technical, ~2 M in THF/heptane/ethylbenzene

Synonym(s):

LDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2NLi
CAS Number:
Molecular Weight:
107.12
Beilstein:
3655042
MDL number:
UNSPSC Code:
12352000

grade

technical

form

liquid

concentration

~2 M in THF/heptane/ethylbenzene

color

brown

density

0.81 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

[Li]N(C(C)C)C(C)C

InChI

1S/C6H14N.Li/c1-5(2)7-6(3)4;/h5-6H,1-4H3;/q-1;+1

InChI key

ZCSHNCUQKCANBX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

may contain precipitate

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service