Skip to Content
Merck
All Photos(1)

Key Documents

53422

Sigma-Aldrich

HMPA-(aminomethyl)polystyrene

extent of labeling: ~1.1 mmol/g loading

Synonym(s):

(4-Hydroxymethyl-3-methoxyphenoxy)acetyl-AM-polystyrene, HMPA resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

extent of labeling

~1.1 mmol/g loading

matrix

crosslinked with 1% DVB

particle size

200-400 mesh

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service